Learn More
Medchemexpress LLC C2/Complement C2 Mo 500ug
SDP
Catalog No. 5000178881
Encompass_Preferred
Complement C2 is a component of the classical pathway of the complement system and is cleaved by activated factor C1 resulting in the formation of two distinct fragments C2b and C2a The subsequent serine protease activity of C2a is critical for its subsequent interaction with the complement factor C4b leading to the formation of C3 or C5 convertase C2/Complement C2 Protein Mouse (HEK293 His) is the recombinant mouse-derived C2/Complement C2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78881 Supplier Medchemexpress LLC Supplier No. HYP74374500UG
Complement C2 is a component of the classical pathway of the complement system and is cleaved by activated factor C1 resulting in the formation of two distinct fragments C2b and C2a The subsequent serine protease activity of C2a is critical for its subsequent interaction with the complement factor C4b leading to the formation of C3 or C5 convertase C2/Complement C2 Protein Mouse (HEK293 His) is the recombinant mouse-derived C2/Complement C2 protein expressed by HEK293 with C-His labeled tag