Learn More
Enzo Life Sciences C2 Dihydroceramide (5 mg). CAS: 13031-64-6
SDP
Supplier:  Enzo Life Sciences BMLSL1010005
Encompass_Preferred
C2 Dihydroceramide is inactive and may be used as a negative control for C2 ceramide. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (5mg/ml) or DMSO (5mg/ml: dissolve in hot DMSO, then cool to RT). Long Term Storage: -20°C.
Catalog No. 50-201-2102