Learn More
Cayman Chemical C2 L-threo Ceramide (d18:1/2:0), Purity: ≥98%, CAS: 143615-69-4, 1 mg
SDP
Catalog No. 502933858
Encompass_Preferred
A cell-permeable analog of naturally occurring ceramides; stimulates cholesterol efflux in CHO cells expressing the human ABCA1 receptor (10 μM); inhibits IL-4 production by 17% in EL4 T cells stimulated with PMA when used at a concentration of 10 μM; induces cell cycle arrest in the G0/G1 phase and a 7-fold increase in sphingosine accumulation as well as inhibits growth of HL-60 leukemia cells
Catalog No. 50-293-3858 Supplier Cayman Chemical Supplier No. 243861 MG
A cell-permeable analog of naturally occurring ceramides; stimulates cholesterol efflux in CHO cells expressing the human ABCA1 receptor (10 μM); inhibits IL-4 production by 17% in EL4 T cells stimulated with PMA when used at a concentration of 10 μM; induces cell cycle arrest in the G0/G1 phase and a 7-fold increase in sphingosine accumulation as well as inhibits growth of HL-60 leukemia cells