Learn More
Enzo Life Sciences C2-Phytoceramide (5 mg). CAS: 475995-69-8
SDP
Supplier:  Enzo Life Sciences BMLSL1510005
Encompass_Preferred
Inhibits yeast growth and activates ceramide-activated protein phosphatase (CAPP). Purity: ≥98% (TLC). Solubility: Soluble in ethanol (>25 mg/ml) or DMSO (>25 mg/ml with warming). Long Term Storage: -20°C.
Catalog No. 50-201-2124