Learn More
Sorbent Technologies Inc C2 Silica TLC Plates, w/UV254, glass backed, 250um, 20x20cm, 25/pk
SDP
Catalog No. 501138617
Encompass_Preferred
This thin layer chromatography (TLC) plate comprises a C2 Silica layer. It can be developed with purely organic, organic/aqueous and purely aqueous eluents. Thus the phase finds numerous applications in normal phase chromatography as well as in the reversed phase mode. The relative polarity of the eluent determines the polarity of the layer. The Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 5-17um, and pH stability of 2-10. The TLC plates contain UV254 fluorescent indicator. The UV-absorbing substances appear as dark-blue to black spots on a light - blue background. This analytical layer is bound to a glass support. It has a thickness of 250um and dimensions of 20x20cm. There are 25 plates per pack. Applications include: active plant constituents, steroids
Catalog No. 50-113-8617 Supplier Sorbent Technologies Inc Supplier No. 2815126
This thin layer chromatography (TLC) plate comprises a C2 Silica layer. It can be developed with purely organic, organic/aqueous and purely aqueous eluents. Thus the phase finds numerous applications in normal phase chromatography as well as in the reversed phase mode. The relative polarity of the eluent determines the polarity of the layer. The Silica 60 has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, particle size of 5-17um, and pH stability of 2-10. The TLC plates contain UV254 fluorescent indicator. The UV-absorbing substances appear as dark-blue to black spots on a light - blue background. This analytical layer is bound to a glass support. It has a thickness of 250um and dimensions of 20x20cm. There are 25 plates per pack. Applications include: active plant constituents, steroids