Learn More
Cayman Chemical C20 Ceramide (d18:1/20:0), Purity: ≥98%, CAS: 7344-02-7, 1 mg
SDP
Catalog No. 502887353
Encompass_Preferred
A natural 20:0 ceramide that is abundant in the brain; synthesized de novo by ceramide synthases 1 and 2
Catalog No. 50-288-7353 Supplier Cayman Chemical Supplier No. 107241 MG
A natural 20:0 ceramide that is abundant in the brain; synthesized de novo by ceramide synthases 1 and 2