Learn More
Medchemexpress LLC C20 Ceramide (N-Arachidoyl-D-sphingosine) | 7344-02-7 | 99.1% | 594.01 | 100 MG
SDP
Catalog No. 5000438997
Encompass_Preferred
C20 Ceramide (N-Arachidoyl-D-sphingosine) is a natural 20:0 ceramide and a N-acylsphingosine with an eicosanoyl N-acyl group. It functions as a crucial signal molecule in various biological processes, including cell growth, differentiation, senescence, and apoptosis.
  • Natural 20:0 ceramide
  • N-acylsphingosine with an eicosanoyl N-acyl group
  • Mediates biological processes
  • Involved in cell growth, differentiation, senescence, and apoptosis
Catalog No. 50-004-38997 Supplier Medchemexpress LLC Supplier No. HY112016100MG
C20 Ceramide (N-Arachidoyl-D-sphingosine) is a natural 20:0 ceramide and a N-acylsphingosine with an eicosanoyl N-acyl group. It functions as a crucial signal molecule in various biological processes, including cell growth, differentiation, senescence, and apoptosis.
  • Natural 20:0 ceramide
  • N-acylsphingosine with an eicosanoyl N-acyl group
  • Mediates biological processes
  • Involved in cell growth, differentiation, senescence, and apoptosis