Learn More
Medchemexpress LLC C20 Ceramide (N-Arachidoyl-D-sphingosine) | 7344-02-7 | 99.1% | 594.01 | 25 MG
SDP
Catalog No. 5000439518
Encompass_Preferred
C20 Ceramide (N-Arachidoyl-D-sphingosine) is a natural 20:0 ceramide and a N-acylsphingosine in which the ceramide N-acyl group is specified as eicosanoyl. C20 Ceramide acts as a key signal molecule mediating biological processes. It is for research use only and not sold to patients.
  • Natural 20:0 ceramide
  • N-acylsphingosine with eicosanoyl N-acyl group
  • Acts as a key signal molecule
  • Mediates cell growth, differentiation, senescence, and apoptosis
  • For research use only
Catalog No. 50-004-39518 Supplier Medchemexpress LLC Supplier No. 25MGHY112016
C20 Ceramide (N-Arachidoyl-D-sphingosine) is a natural 20:0 ceramide and a N-acylsphingosine in which the ceramide N-acyl group is specified as eicosanoyl. C20 Ceramide acts as a key signal molecule mediating biological processes. It is for research use only and not sold to patients.
  • Natural 20:0 ceramide
  • N-acylsphingosine with eicosanoyl N-acyl group
  • Acts as a key signal molecule
  • Mediates cell growth, differentiation, senescence, and apoptosis
  • For research use only