Learn More
Cayman Chemical C20 nd C22 Faty ACD Methyl 1mL
SDP
Catalog No. 502988124
Encompass_Preferred
A mixture of C20 and C22 fatty acid methyl esters; contains six C20 and one C22 fatty acid methyl esters; intended for use as a reference standard for the detection of C20 and C22 fatty acids in various sample types by GC-MS
Catalog No. 50-298-8124 Supplier Cayman Chemical Supplier No. 293361 ML
A mixture of C20 and C22 fatty acid methyl esters; contains six C20 and one C22 fatty acid methyl esters; intended for use as a reference standard for the detection of C20 and C22 fatty acids in various sample types by GC-MS