Learn More
Aviva Systems Biology Corp C20ORF100 Antibody - N-terminal region (ARP30008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20ORF100. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human C20ORF100. Alternate names: GCX-1, C20orf100, dJ495O3.1, dJ1108D11.2ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52815