Learn More
Aviva Systems Biology Corp C20orf103 Antibody - middle region (ARP46875_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf103. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C20orf103. Alternate names: LAMP-5, UNC-43, BAD-LAMP, C20orf103ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53496