Learn More
Aviva Systems Biology Corp C20orf160 Antibody - N-terminal region (ARP52506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf160. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C20orf160. Alternate names: FLJ43600, dJ310O13.5, C20orf160ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60756