Learn More
Aviva Systems Biology Corp C20orf18 Antibody - middle region (ARP43073_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43073_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C20orf18. Alternate names: RP5-852M4.4, C20orf18, HOIL1, RBCK2, RNF54, UBCE7IP3, XAP4, ZRANB4, XAP3, HOIL-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58416