Learn More
Aviva Systems Biology Corp C20ORF20 Antibody - N-terminal region (ARP33351_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33351_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C20ORF20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C20ORF20. Alternate names: Eaf7, MRGBP, URCC4, MRG15BP, C20ORF20ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54975