Learn More
Aviva Systems Biology Corp RAB5IF Antibody - N-terminal region (ARP49339_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49339_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C20orf24. Alternate names: PNAS-11, RIP5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-60066