Learn More
Aviva Systems Biology Corp C20orf30 Antibody - C-terminal region (ARP44585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C20orf30. Alternate names: HSPC274, dJ1116H23.2.1, C20orf30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58852