Learn More
Medchemexpress LLC C21H15ClNNa2O4PS | 193884-53-6 | 98.0% | 489.82 | 250 MG
SDP
Catalog No. 5000439485
Encompass_Preferred
Lumigen APS-5 is a substrate of alkaline phosphatase (ALP). It can be used to assess the activity of alkaline phosphatase (ALP).
  • Used to assess the activity of alkaline phosphatase (ALP).
  • For research use only.
Catalog No. 50-004-39485 Supplier Medchemexpress LLC Supplier No. 250MGHY112832
Lumigen APS-5 is a substrate of alkaline phosphatase (ALP). It can be used to assess the activity of alkaline phosphatase (ALP).
  • Used to assess the activity of alkaline phosphatase (ALP).
  • For research use only.