Learn More
Medchemexpress LLC C21H18F7N3O4 | 98.0% | 509.37 | 1 MG
SDP
Catalog No. 5000392896
Encompass_Preferred
RTx-161 is an allosteric Polθ polymerase inhibitor with an IC50 value of 4.1 nM. It selectively kills HR-deficient cancer cells and suppresses PARP inhibitor (PARPi) resistance in multiple genetic backgrounds, including HR-proficient cells. RTx-161 can also induce apoptosis.
  • Allosteric Polθ polymerase inhibitor with an IC50 of 4.1 nM.
  • Selectively kills HR-deficient cancer cells.
  • Suppresses PARP inhibitor (PARPi) resistance in multiple genetic backgrounds, including HR-proficient cells.
  • Can induce apoptosis.
Catalog No. 50-003-92896 Supplier Medchemexpress LLC Supplier No. HY1614301MG
RTx-161 is an allosteric Polθ polymerase inhibitor with an IC50 value of 4.1 nM. It selectively kills HR-deficient cancer cells and suppresses PARP inhibitor (PARPi) resistance in multiple genetic backgrounds, including HR-proficient cells. RTx-161 can also induce apoptosis.
  • Allosteric Polθ polymerase inhibitor with an IC50 of 4.1 nM.
  • Selectively kills HR-deficient cancer cells.
  • Suppresses PARP inhibitor (PARPi) resistance in multiple genetic backgrounds, including HR-proficient cells.
  • Can induce apoptosis.