Learn More
Aviva Systems Biology Corp C21orf13 Antibody - N-terminal region (ARP45912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C21orf13. Alternate names: MGC33295, C21orf13ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-59165