Learn More
Aviva Systems Biology Corp C21ORF18 Antibody - C-terminal region (ARP34564_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34564_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21ORF18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C21ORF18. Alternate names: C21orf18, C21orf27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55454