Learn More
Aviva Systems Biology Corp C21orf45 Antibody - N-terminal region (ARP45864_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45864_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf45. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C21orf45. Alternate names: B28, C21orf46, FASP1, MIS18alpha, C21orf45, hMis18alphaApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-014-54004