Learn More
Aviva Systems Biology Corp C21orf56 Antibody - middle region (ARP45879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf56. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C21orf56. Alternate names: DKFZp434N0650, MGC99490, C21orf56ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59161