Learn More
Aviva Systems Biology Corp C21orf62 Antibody - middle region (ARP45868_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45868_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf62. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C21orf62. Alternate names: B37, C21orf120, PRED81ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-59157