Learn More
Aviva Systems Biology Corp C21orf7 Antibody - C-terminal region (ARP42552_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42552_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human C21orf7. Alternate names: TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4, HC21ORF7, MAP3K7CLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58262