Learn More
Aviva Systems Biology Corp C21orf87 Antibody - C-terminal region (ARP45782_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45782_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf87. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C21orf87. Alternate names: C21orf87, NCRNA00257ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59137