Learn More
Aviva Systems Biology Corp C21orf91 Antibody - middle region (ARP45860_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45860_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf91. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C21orf91. Alternate names: C21orf14, C21orf38, DKFZp781D1223, YG81, EURL, CSSG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59155