Learn More
Medchemexpress LLC C22-Ceramide | 27888-44-4 | MFCD09840213 | C40H79NO3 | 25 MG
SDP
Catalog No. 5000375077
Encompass_Preferred
C22-Ceramide is an endogenous bioactive sphingolipid. It reduces the propensity of C16-ceramide channel formation in isolated rat liver mitochondria and in liposomes, making it suitable for various research applications.
  • Endogenous bioactive sphingolipid
  • Reduces C16-ceramide channel formation in isolated rat liver mitochondria
  • Reduces C16-ceramide channel formation in liposomes
  • High purity of 99.89%
  • Supplied as a white to off-white solid
  • Suitable for research use only
  • Stable for shipping at room temperature (continental US)
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
Catalog No. 50-003-75077 Supplier Medchemexpress LLC Supplier No. 25MGHY154831
C22-Ceramide is an endogenous bioactive sphingolipid. It reduces the propensity of C16-ceramide channel formation in isolated rat liver mitochondria and in liposomes, making it suitable for various research applications.
  • Endogenous bioactive sphingolipid
  • Reduces C16-ceramide channel formation in isolated rat liver mitochondria
  • Reduces C16-ceramide channel formation in liposomes
  • High purity of 99.89%
  • Supplied as a white to off-white solid
  • Suitable for research use only
  • Stable for shipping at room temperature (continental US)
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)