Learn More
Cayman Chemical C22 CeramIded18 122 0 10mg
SDP
Catalog No. 502987736
Encompass_Preferred
A bioactive sphingolipid; forms small channels in liposomes, suggesting that acyl chain length is important in mitochondrial-mediated apoptosis; decreases the mitochondrial-to-cytosolic stress response in C. elegans following hsp-6 RNAi
Catalog No. 50-298-7736 Supplier Cayman Chemical Supplier No. 2253310 MG
A bioactive sphingolipid; forms small channels in liposomes, suggesting that acyl chain length is important in mitochondrial-mediated apoptosis; decreases the mitochondrial-to-cytosolic stress response in C. elegans following hsp-6 RNAi