Learn More
Cayman Chemical C22 Ceramide (d18:1/22:0), Purity: ≥95%, CAS: 27888-44-4, 5 mg
SDP
Catalog No. 502986727
Encompass_Preferred
A bioactive sphingolipid; forms small channels in liposomes, suggesting that acyl chain length is important in mitochondrial-mediated apoptosis; decreases the mitochondrial-to-cytosolic stress response in C. elegans following hsp-6 RNAi
Catalog No. 50-298-6727 Supplier Cayman Chemical Supplier No. 225335 MG
A bioactive sphingolipid; forms small channels in liposomes, suggesting that acyl chain length is important in mitochondrial-mediated apoptosis; decreases the mitochondrial-to-cytosolic stress response in C. elegans following hsp-6 RNAi