Learn More
Cayman Chemical C22 Glucosylceramide-d4 (d18:1/22:0-d4), Purity: ≥99% deuterated forms (d1-d4), CAS: 2738376-81-1, 1 mg
SDP
Catalog No. 502895400
Encompass_Preferred
An internal standard for the quantification of C22 glucosylceramide by GC- or LC-MS
Catalog No. 50-289-5400 Supplier Cayman Chemical Supplier No. 90034621 MG
An internal standard for the quantification of C22 glucosylceramide by GC- or LC-MS