Learn More
Alkali Scientific Alkali Scientific™ C2248 StripSpin™ 12D Digital Mini Centrifuge w/ PCR Strip Rotor
SDP
Catalog No. 503029675
Encompass_Preferred
The new Alkali Scientiifc C2248 StripSpin 12D digital mini centrifuge from Alkali is uniquely designed for centrifugation of 12 position PCR strips. It features digital speed control up to 5,500 rpm, timer control up to 99 min., and a safety interlock to stop the centrifuge if the lid is opened during operation. With built in rcf conversion, speed can be set/displayed in rpm or rcf, the Alkali Scientific™ C2248 StripSpin 12D is the perfect solution for centrifugation of up to 4 x 12-position high efficiency PCR strips (or 48 x 0.2ml tubes) and includes a 2 year warranty.
Catalog No. 50-302-9675 Supplier Alkali Scientific Supplier No. RS7186
The new Alkali Scientiifc C2248 StripSpin 12D digital mini centrifuge from Alkali is uniquely designed for centrifugation of 12 position PCR strips. It features digital speed control up to 5,500 rpm, timer control up to 99 min., and a safety interlock to stop the centrifuge if the lid is opened during operation. With built in rcf conversion, speed can be set/displayed in rpm or rcf, the Alkali Scientific™ C2248 StripSpin 12D is the perfect solution for centrifugation of up to 4 x 12-position high efficiency PCR strips (or 48 x 0.2ml tubes) and includes a 2 year warranty.