Learn More
Aviva Systems Biology Corp C22orf28 Antibody - N-terminal region (ARP56745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C22orf28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C22orf28. Alternate names: DJ149A16.6, HSPC117, RP1-149A16.6, C22orf28ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61971