Learn More
Aviva Systems Biology Corp C22orf30 Antibody - middle region (ARP55650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C22orf30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C22orf30. Alternate names: DKFZp781P2328, FLJ23059, MGC50372, C22orf30, RP4-694E4.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61565