Learn More
Aviva Systems Biology Corp LRRC75B Antibody - middle region (ARP56011_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C22orf36. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human C22orf36. Alternate names: MGC131773, C22orf36ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-286-107