Learn More
Cayman Chemical C23 Globotriaosylceramide d1
SDP
Catalog No. 503015990
Encompass_Preferred
A sphingolipid; has been used as a standard for the quantification of globotriaosylceramide in human fibroblasts by HPLC/APCI-MS
Catalog No. 50-301-5990 Supplier Cayman Chemical Supplier No. 24878500 UG
A sphingolipid; has been used as a standard for the quantification of globotriaosylceramide in human fibroblasts by HPLC/APCI-MS