Learn More
Cayman Chemical C24 1 1-DeoxyceramIdem18 1 1mg
SDP
Catalog No. 502988145
Encompass_Preferred
A very long-chain atypical ceramide containing a 1-deoxysphingosine (m18:1(4E)) backbone
Catalog No. 50-298-8145 Supplier Cayman Chemical Supplier No. 300301 MG
A very long-chain atypical ceramide containing a 1-deoxysphingosine (m18:1(4E)) backbone