Learn More
Cayman Chemical C24 1 3sulfo GalactosylcRa 1mg
SDP
Catalog No. 502934709
Encompass_Preferred
A sulfatide; interacts with LMIR5 to increase activation of NFAT; an immunodominant species that is bound by CD1d in vitro; reduces symptoms, increases survival, and decreases inflammatory lesions in a mouse model of chronic relapsing-remitting EAE (20 UG)
Catalog No. 50-293-4709 Supplier Cayman Chemical Supplier No. 248651 MG
A sulfatide; interacts with LMIR5 to increase activation of NFAT; an immunodominant species that is bound by CD1d in vitro; reduces symptoms, increases survival, and decreases inflammatory lesions in a mouse model of chronic relapsing-remitting EAE (20 UG)