Learn More
Medchemexpress LLC C24:1 ceramide (d18:1/24:1(15Z)) | 54164-50-0 | MFCD02259277 | 99.9% | C42H81NO3 | 10MG
SDP
Catalog No. 5000259246
Encompass_Preferred
C24:1 ceramide (d18:1/24:1(15Z)) is a long-chain monounsaturated ceramide supplied as a high-purity research reagent for biochemical and lipidomics studies. It is provided as a solid standard suitable for analytical assays, lipid formulation, and cell signaling research.
  • High purity for reproducible analytical results.
  • Suitable for lipidomics and biochemical assays.
  • Solid standard, easy to weigh and store.
  • Supports studies of cell signaling and apoptosis.
  • Stable under recommended storage conditions.
Catalog No. 50-002-59246 Supplier Medchemexpress LLC Supplier No. HY13457510MG
C24:1 ceramide (d18:1/24:1(15Z)) is a long-chain monounsaturated ceramide supplied as a high-purity research reagent for biochemical and lipidomics studies. It is provided as a solid standard suitable for analytical assays, lipid formulation, and cell signaling research.
  • High purity for reproducible analytical results.
  • Suitable for lipidomics and biochemical assays.
  • Solid standard, easy to weigh and store.
  • Supports studies of cell signaling and apoptosis.
  • Stable under recommended storage conditions.