Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C24:1-ceramide (d18:1/24:1) | 54164-50-0 | MFCD02259277 | 99.9% | 648.10 g·mol⁻¹ | C42H81NO3 | 5 MG
SDP

Catalog No. 5000222039
Encompass_Preferred

C24:1-Ceramide (d18:1/24:1) is a long-chain sphingolipid commonly used in membrane biology, lipidomics, and formulation research. It functions as a reference standard and a structural lipid in liposome and lipid nanoparticle preparations for in vitro and analytical studies.

  • High chemical purity reported by distributors (approximately 99.9%).
  • Molecular formula C42H81NO3 and molecular weight ~648.10 g·mol-1.
  • Solid form provided in small research pack sizes (mg scale) for analytical use.
  • Suitable for membrane and lipidomics studies, and lipid nanoparticle formulation.
  • Recommended storage at -20 °C to preserve stability.
  • Supplied with CAS identifier 54164-50-0 for unambiguous reference.

Catalog No. 50-002-22039 Supplier Medchemexpress LLC Supplier No. HY1345755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C24:1-Ceramide (d18:1/24:1) is a long-chain sphingolipid commonly used in membrane biology, lipidomics, and formulation research. It functions as a reference standard and a structural lipid in liposome and lipid nanoparticle preparations for in vitro and analytical studies.

  • High chemical purity reported by distributors (approximately 99.9%).
  • Molecular formula C42H81NO3 and molecular weight ~648.10 g·mol-1.
  • Solid form provided in small research pack sizes (mg scale) for analytical use.
  • Suitable for membrane and lipidomics studies, and lipid nanoparticle formulation.
  • Recommended storage at -20 °C to preserve stability.
  • Supplied with CAS identifier 54164-50-0 for unambiguous reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.