Learn More
Medchemexpress LLC C24:1 dihydro 1-deoxyceramide (m18:0/24:1) | 1246298-60-1 | MFCD22416693 | 99.0% | 634.11 g/mol | C42H83NO2 | 5 MG
SDP
Catalog No. 5000222205
Encompass_Preferred
C24:1 dihydro 1-deoxyceramide (m18:0/24:1) is a long-chain deoxyceramide provided for research use. Supplied as a white to off-white solid, it is used as a reference standard or reagent in studies of sphingolipid metabolism, lipidomics, and biochemical assays.
  • High purity (99.0%).
  • White to off-white solid appearance.
  • Suitable for lipidomics and sphingolipid research applications.
  • Provided with COA, SDS, and data sheet available.
  • Offered in small milligram pack sizes for analytical work.
Catalog No. 50-002-22205 Supplier Medchemexpress LLC Supplier No. HY1392845MG
C24:1 dihydro 1-deoxyceramide (m18:0/24:1) is a long-chain deoxyceramide provided for research use. Supplied as a white to off-white solid, it is used as a reference standard or reagent in studies of sphingolipid metabolism, lipidomics, and biochemical assays.
  • High purity (99.0%).
  • White to off-white solid appearance.
  • Suitable for lipidomics and sphingolipid research applications.
  • Provided with COA, SDS, and data sheet available.
  • Offered in small milligram pack sizes for analytical work.