Learn More
Cayman Chemical C24 1 dIhydro 1DeoxycRamID 1mg
SDP
Catalog No. 502937314
Encompass_Preferred
A very long-chain atypical ceramide containing a 1-deoxysphinanine (m18:0) backbone; has been found as a major 1-deoxyceramide species in MEFs following application of 1-deoxysphinganine alkyne or 1-deoxysphinanine-d3
Catalog No. 50-293-7314 Supplier Cayman Chemical Supplier No. 275621 MG
A very long-chain atypical ceramide containing a 1-deoxysphinanine (m18:0) backbone; has been found as a major 1-deoxyceramide species in MEFs following application of 1-deoxysphinganine alkyne or 1-deoxysphinanine-d3