Learn More
Cayman Chemical C24 CeramIded18 1 14Z24 0 1mg
SDP
Catalog No. 502938804
Encompass_Preferred
A long-chain atypical ceramide; contains a sphingosine (d18:1(14Z)) backbone with a cis double bond at the 14-15 position
Catalog No. 50-293-8804 Supplier Cayman Chemical Supplier No. 286041 MG
A long-chain atypical ceramide; contains a sphingosine (d18:1(14Z)) backbone with a cis double bond at the 14-15 position