Learn More
Strem, An Ascensus Company CAS# 53199-31-8. Bis(tri-t-butylphosphine)palladium(0), 98%. MFCD03094580. 25G
SDP
Catalog No. 501959494
Encompass
CAS# 53199-31-8. 25G. Bis(tri-t-butylphosphine)palladium(0), 98%. MFCD03094580. Molecular Formula: C24H54P2Pd. Molecular Weight: 511.06. Color/Form: off-white xtl. Stability: air sensitive. Strem# 46-0252. www.strem.com/catalog/v3/46-0252/
Catalog No. 50-195-9494 Supplier Strem, An Ascensus Company Supplier No. 46025225G
CAS# 53199-31-8. 25G. Bis(tri-t-butylphosphine)palladium(0), 98%. MFCD03094580. Molecular Formula: C24H54P2Pd. Molecular Weight: 511.06. Color/Form: off-white xtl. Stability: air sensitive. Strem# 46-0252. www.strem.com/catalog/v3/46-0252/