Learn More
Alkali Scientific C2595 MyFuge 5D Digital Micro
SDP
Catalog No. 503029671
Encompass_Preferred
The myFuge5D digital mini centrifuge includes a four position rotor that accepts a variety of 5ml tubes (including 5ml snap-cap and screw-cap centrifuge tubes as well as 5ml blood collection tubes) In addition the rotor has four positions for positions for standard microtubes (1 5/2 0ml) and offers optional adapters for 2ml cryotubes 0 2ml PCR tubes and 0 5ml microtubes making it one of the most versatile and compact centrifuges in its class The myFuge 5D offers digital speed control (up to 5500 rpm) Timer is also controlled (up to 99 min ) with a safety interlock to stop the centrifuge if the lid is opened during operation With built in rcf conversion speed can be set/displayed in rpm or rcf The myFuge 5D also comes with a 2-year warranty HEPA Filter and UV light reduce contamination Highly compact 22 x 16in footprint Included cable access port allows use of centrifuges vortexers and other instruments Included pipette holder and shelf provides storage for pipettes and tips
Catalog No. 50-302-9671 Supplier Alkali Scientific Supplier No. RS2205
The myFuge5D digital mini centrifuge includes a four position rotor that accepts a variety of 5ml tubes (including 5ml snap-cap and screw-cap centrifuge tubes as well as 5ml blood collection tubes) In addition the rotor has four positions for positions for standard microtubes (1 5/2 0ml) and offers optional adapters for 2ml cryotubes 0 2ml PCR tubes and 0 5ml microtubes making it one of the most versatile and compact centrifuges in its class The myFuge 5D offers digital speed control (up to 5500 rpm) Timer is also controlled (up to 99 min ) with a safety interlock to stop the centrifuge if the lid is opened during operation With built in rcf conversion speed can be set/displayed in rpm or rcf The myFuge 5D also comes with a 2-year warranty HEPA Filter and UV light reduce contamination Highly compact 22 x 16in footprint Included cable access port allows use of centrifuges vortexers and other instruments Included pipette holder and shelf provides storage for pipettes and tips