Learn More
Cayman Chemical C26 1 Sphingomyelin-d5 d18 1
SDP
Catalog No. 502896023
Encompass_Preferred
An internal standard for the quantification of C26:1 sphingomyelin by GC- or LC-MS
Catalog No. 50-289-6023 Supplier Cayman Chemical Supplier No. 9003463500 UG
An internal standard for the quantification of C26:1 sphingomyelin by GC- or LC-MS