Learn More
Medchemexpress LLC C26 ceramide | 121459-09-4 | 99.9% | 678.17 g/mol | C44H87NO3 | 10 MG
SDP
Catalog No. 5000284507
Encompass_Preferred
C26-Ceramide is a long-chain ceramide lipid (N-hexacosanoylsphingosine) composed of one saturated and one unsaturated hydrophobic tail. It is provided as a high-purity biochemical reagent for lipid research and formulation studies, including lipid nanoparticle and liposome development.
  • High purity (99.89%) suitable for research applications.
  • Long-chain ceramide structure with a C26 acyl chain.
  • Useful for lipid nanoparticle and liposome formulation studies.
  • Supplied in research-scale quantities for assay and formulation work.
  • Molecular formula C44H87NO3; molecular weight 678.17 g/mol.
Catalog No. 50-002-84507 Supplier Medchemexpress LLC Supplier No. HYW59067410MG
C26-Ceramide is a long-chain ceramide lipid (N-hexacosanoylsphingosine) composed of one saturated and one unsaturated hydrophobic tail. It is provided as a high-purity biochemical reagent for lipid research and formulation studies, including lipid nanoparticle and liposome development.
  • High purity (99.89%) suitable for research applications.
  • Long-chain ceramide structure with a C26 acyl chain.
  • Useful for lipid nanoparticle and liposome formulation studies.
  • Supplied in research-scale quantities for assay and formulation work.
  • Molecular formula C44H87NO3; molecular weight 678.17 g/mol.