Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C26-ceramide | 121459-09-4 | MFCD18379264 | 99.9% | 678.17 g/mol | C44H87NO3 | 1 MG
SDP

Catalog No. 5000284267
Encompass_Preferred

C26-Ceramide is a very long-chain ceramide lipid consisting of a sphingoid base N-acylated with a C26 fatty acid. It is supplied as a high-purity biochemical reagent for lipid research and formulation work, including development of lipid nanoparticles and liposomes.

  • Very long-chain ceramide with one saturated and one unsaturated hydrophobic tail.
  • High purity (approximately 99.9%) suitable for analytical and formulation studies.
  • Molecular weight 678.17 g/mol; chemical formula C44H87NO3.
  • Supplied in small research quantities (example: 1 mg) for experimental use.
  • Stable as a powder when stored at -20 °C; follow laboratory handling and storage guidelines.

Catalog No. 50-002-84267 Supplier Medchemexpress LLC Supplier No. HYW5906741MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C26-Ceramide is a very long-chain ceramide lipid consisting of a sphingoid base N-acylated with a C26 fatty acid. It is supplied as a high-purity biochemical reagent for lipid research and formulation work, including development of lipid nanoparticles and liposomes.

  • Very long-chain ceramide with one saturated and one unsaturated hydrophobic tail.
  • High purity (approximately 99.9%) suitable for analytical and formulation studies.
  • Molecular weight 678.17 g/mol; chemical formula C44H87NO3.
  • Supplied in small research quantities (example: 1 mg) for experimental use.
  • Stable as a powder when stored at -20 °C; follow laboratory handling and storage guidelines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.