Learn More
Sigma Aldrich Fine Chemicals Biosciences Cytochalasin D, 22144-77-0, MFCD00077706, 200 ul
Catalog No. NC1118425
Encompass
Empirical Formula (Hill Notation): C30H37NO6, Molecular Weight: 507.62, Synonym: Zygosporin A, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO, 0.2 um filtered. Cytochalasin B is used to inhibit sugar transport in fibroblasts , decrease cytoskeletal actin and myosin in rabbit neutrophils , and to stimulate B lymphocytes . It is a potent inhibitor of actin polymerization, disrupts actin microfilaments, activates the p53-dependent pathways, inhibits smooth muscle contraction, and inhibits insulin-stimulated glucose transport.
Catalog No. NC1118425 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C2618200UL
Empirical Formula (Hill Notation): C30H37NO6, Molecular Weight: 507.62, Synonym: Zygosporin A, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO, 0.2 um filtered. Cytochalasin B is used to inhibit sugar transport in fibroblasts , decrease cytoskeletal actin and myosin in rabbit neutrophils , and to stimulate B lymphocytes . It is a potent inhibitor of actin polymerization, disrupts actin microfilaments, activates the p53-dependent pathways, inhibits smooth muscle contraction, and inhibits insulin-stimulated glucose transport.