Learn More
Medchemexpress LLC C26H19F5N6OS | 1448316-08-2 | MFCD31657322 | 99.9% | 558.53 | C26H19F5N6OS | 5 MG
SDP
Catalog No. 5000213255
Encompass_Preferred
BAY-826 is a small-molecule chemical probe that selectively and potently inhibits the TIE-2 (TEK) receptor tyrosine kinase. It demonstrates high biochemical affinity (Kd ≈ 1.6 nM) and potent cellular activity (EC50 ≈ 1.3 nM for inhibition of TIE-2 autophosphorylation in endothelial cells). Intended for research use, the compound is supplied in multiple pack sizes and in DMSO solutions for in vitro studies.
  • Selective TIE-2 inhibition with low-nanomolar affinity.
  • Potent cellular activity against TIE-2 autophosphorylation.
  • High reported purity suitable for biochemical assays.
  • Available in multiple small-scale and bulk pack sizes.
  • Provided as solid and as DMSO solution formats for convenience.
Catalog No. 50-002-13255 Supplier Medchemexpress LLC Supplier No. HY1007565MG
BAY-826 is a small-molecule chemical probe that selectively and potently inhibits the TIE-2 (TEK) receptor tyrosine kinase. It demonstrates high biochemical affinity (Kd ≈ 1.6 nM) and potent cellular activity (EC50 ≈ 1.3 nM for inhibition of TIE-2 autophosphorylation in endothelial cells). Intended for research use, the compound is supplied in multiple pack sizes and in DMSO solutions for in vitro studies.
  • Selective TIE-2 inhibition with low-nanomolar affinity.
  • Potent cellular activity against TIE-2 autophosphorylation.
  • High reported purity suitable for biochemical assays.
  • Available in multiple small-scale and bulk pack sizes.
  • Provided as solid and as DMSO solution formats for convenience.