Learn More
New Jersey Fluid Systems LLC HC-400-7-4 Hastelloy C-276 Female Connector, Leak-Tight Gas Seal, Excellent Corrosion Resistance, 1/4 in
Catalog No. NC3714511
Encompass
The HC-400-7-4 Hastelloy C-276 Female Connector is engineered for optimal performance in critical fluid systems. Its two-ferrule design ensures reliable, leak-tight connections, exceptional corrosion resistance, and high-temperature capability, making it ideal for demanding applications. Manufactured by Swagelok, this connector excels in preventing corrosion and offers easy installation without torque transmission.
Catalog No. NC3714511 Supplier New Jersey Fluid Systems LLC Supplier No. HC40074
The HC-400-7-4 Hastelloy C-276 Female Connector is engineered for optimal performance in critical fluid systems. Its two-ferrule design ensures reliable, leak-tight connections, exceptional corrosion resistance, and high-temperature capability, making it ideal for demanding applications. Manufactured by Swagelok, this connector excels in preventing corrosion and offers easy installation without torque transmission.